2-[2-(tert-Butyldimethylsiloxy)ethyl]benzeneboronic acid 96%

Supplier: Apollo Scientific
Warning

OR3328-1G OR3328-250MG
APOSOR3328-1GEA 596 EUR
APOSOR3328-1G APOSOR3328-250MG
2-[2-(tert-Butyldimethylsiloxy)ethyl]benzeneboronic acid 96%
2-[2-(tert-Butyldimethylsiloxy)ethyl]benzeneboronic acid
Formula: C₁₄H₂₅BO₃Si
MW: 280,25 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Freezer
MDL Number: MFCD08436049
CAS Number: 913835-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR