1-Methoxy-2-(1H-pyrrol-1-yl)ethyl methyl ether 95%
Supplier: Apollo Scientific
Synonyms:
1-(2,2-Dimethoxyethyl)-1H-pyrrole
OR33312-1G
OR33312-5G
APOSOR33312-1GEA
180
EUR
APOSOR33312-1G
APOSOR33312-5G
1-Methoxy-2-(1H-pyrrol-1-yl)ethyl methyl ether 95%
1-Methoxy-2-(1H-pyrrol-1-yl)ethyl methyl ether
Formula:
C₈H₁₃NO₂ MW: 155,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659604 CAS Number: 93217-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...