2-Chloro-N-(4-chlorobenzyl)acetamide 95%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-N-[(4-chlorophenyl)methyl]acetamide

OR33346-1G OR33346-25G OR33346-5G
APOSOR33346-1GEA 180 EUR
APOSOR33346-1G APOSOR33346-25G APOSOR33346-5G
2-Chloro-N-(4-chlorobenzyl)acetamide 95%
2-Chloro-N-(4-chlorobenzyl)acetamide
Formula: C₉H₉Cl₂NO
MW: 218.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00018915
CAS Number: 99585-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR