4-(1H-Imidazol-2-yl)pyridine 95%
Supplier: Apollo Scientific
OR33388-1G
OR33388-5G
APOSOR33388-1GEA
328
EUR
APOSOR33388-1G
APOSOR33388-5G
4-(1H-Imidazol-2-yl)pyridine 95%
4-(1H-Imidazol-2-yl)pyridine
Formula:
C₈H₇N₃ Storage Temperature: Ambient |
MDL Number:
MFCD08668556 CAS Number: 21202-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...