Triisopropyl borate 99%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Tri-i-propylborate
OR3339-1L
OR3339-500ML
OR3339-2.5L
APOSOR3339-1LEA
111
EUR
APOSOR3339-1L
APOSOR3339-500ML
APOSOR3339-2.5L
Triisopropyl borate 99%
Triisopropyl borate
Formula:
C₉H₂₁BO₃ MW: 188,07 g/mol Boiling Pt: 139…141 °C Melting Pt: –59 °C Density: 0,815 g/cm³ (25 °C) Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008872 CAS Number: 5419-55-6 EINECS: 226-529-9 UN: 2616 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...