4-Nitrostyrene 95%

Supplier: Apollo Scientific

OR33416-25G OR33416-5G
APOSOR33416-25GEA 301 EUR
APOSOR33416-25G APOSOR33416-5G
4-Nitrostyrene 95%
4-Nitrostyrene
Formula: C₈H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00041254
CAS Number: 100-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR