7-Acetylbenzoxazole 95%

Supplier: Apollo Scientific

OR33449-1G OR33449-5G
APOSOR33449-1GEA 270 EUR
APOSOR33449-1G APOSOR33449-5G
7-Acetylbenzoxazole 95%
7-Acetylbenzoxazole
Formula: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD09152701
CAS Number: 952182-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR