7-Acetyl-2-butylbenzoxazole 95%

Supplier: Apollo Scientific

Synonyms: 1-(2-Butyl-1,3-benzoxazol-7-yl)ethan-1-one

OR33450-1G OR33450-5G
APOSOR33450-1GEA 270 EUR
APOSOR33450-1G APOSOR33450-5G
7-Acetyl-2-butylbenzoxazole 95%
7-Acetyl-2-butylbenzoxazole
Formula: C₁₃H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD09152702
CAS Number: 952182-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR