7-Acetyl-2-ethylbenzoxazole 95%

Supplier: Apollo Scientific

Synonyms: 1-(2-Ethyl-1,3-benzoxazol-7-yl)ethan-1-one

OR33454-1G OR33454-5G
APOSOR33454-1GEA 270 EUR
APOSOR33454-1G APOSOR33454-5G
7-Acetyl-2-ethylbenzoxazole 95%
7-Acetyl-2-ethylbenzoxazole
Formula: C₁₁H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD09152709
CAS Number: 952182-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR