2-Methoxy-4-morpholinoaniline 95%

Supplier: Apollo Scientific

OR33468-1G OR33468-25G OR33468-5G
APOSOR33468-1GEA 91.6 EUR
APOSOR33468-1G APOSOR33468-25G APOSOR33468-5G
2-Methoxy-4-morpholinoaniline 95%
2-Methoxy-4-morpholinoaniline
Formula: C₁₁H₁₆N₂O₂
MW: 208,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972210
CAS Number: 209960-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR