3-(tert-Butyl)-5-(chloromethyl)-1,2,4-oxadiazole 95%

Supplier: Apollo Scientific

OR33479-1G OR33479-5G
APOSOR33479-1GEA 299 EUR
APOSOR33479-1G APOSOR33479-5G
3-(tert-Butyl)-5-(chloromethyl)-1,2,4-oxadiazole 95%
3-(tert-Butyl)-5-(chloromethyl)-1,2,4-oxadiazole
Formula: C₇H₁₁ClN₂O
MW: 174,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09738869
CAS Number: 944901-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR