6-Hydroxynicotinonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(4-Formylphenyl)isonicotinonitrile, 6-Hydroxypyridine-3-carbonitrile, 5-Cyano-2-hydroxypyridine

OR3348-1G OR3348-250MG
APOSOR3348-1GEA 0 EUR
APOSOR3348-1G APOSOR3348-250MG
6-Hydroxynicotinonitrile 98%
6-Hydroxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Formula: C₆H₄N₂O
Melting Pt: 256…258 °C
Storage Temperature: Ambient
MDL Number: MFCD06656489
CAS Number: 94805-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR