8-Methyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride 95%
Supplier: Apollo Scientific
OR33503-1G
OR33503-5G
APOSOR33503-1GEA
234
EUR
APOSOR33503-1G
APOSOR33503-5G
8-Methyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride 95%
8-Methyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride
Formula:
C₈H₁₇ClN₂S Storage Temperature: Ambient |
MDL Number:
MFCD14584835 CAS Number: 1221575-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...