Ethyl-4-methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate, Technical Grade

Supplier: Apollo Scientific

OR33517-1G OR33517-5G
APOSOR33517-1GEA 336 EUR
APOSOR33517-1G APOSOR33517-5G
Ethyl-4-methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate, Technical Grade
Ethyl-4-methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate
Formula: C₁₂H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD08275093
CAS Number: 54442-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR