Methyl-4-formyl-1H-pyrrole-2-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 4-formyl-1H-pyrrole-2-carboxylate

OR33520-1G OR33520-5G
APOSOR33520-1GEA 68.3 EUR
APOSOR33520-1G APOSOR33520-5G
Methyl-4-formyl-1H-pyrrole-2-carboxylate 95%
Methyl-4-formyl-1H-pyrrole-2-carboxylate
Formula: C₇H₇NO₃
MW: 153,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179575
CAS Number: 40611-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR