Methyl-4-iodo-1H-pyrrole-2-carboxylate 97%

Supplier: Apollo Scientific

Synonyms: Methyl 4-iodopyrrole-2-carboxylate

OR33523-1G OR33523-5G
APOSOR33523-1GEA 270 EUR
APOSOR33523-1G APOSOR33523-5G
Methyl-4-iodo-1H-pyrrole-2-carboxylate 97%
Methyl-4-iodo-1H-pyrrole-2-carboxylate
Formula: C₆H₆INO₂
Storage Temperature: Ambient
MDL Number: MFCD07368824
CAS Number: 40740-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR