3H-Spiro[1,3-benzothiazole-2,4'-oxane] 95%

Supplier: Apollo Scientific

OR33542-1G OR33542-5G
APOSOR33542-1GEA 270 EUR
APOSOR33542-1G APOSOR33542-5G
3H-Spiro[1,3-benzothiazole-2,4'-oxane] 95%
3H-Spiro[1,3-benzothiazole-2,4'-oxane]
Formula: C₁₁H₁₃NOS
Storage Temperature: Ambient
MDL Number: MFCD16140318
CAS Number: 77376-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR