1,3-Thiazolan-2-one 97%
Supplier: Apollo Scientific
Synonyms:
Thiazolidin-2-one
OR33548-100G
OR33548-25G
OR33548-5G
APOSOR33548-100GEA
141
EUR
APOSOR33548-100G
APOSOR33548-25G
APOSOR33548-5G
1,3-Thiazolan-2-one 97%
1,3-Thiazolan-2-one
Formula:
C₃H₅NOS MW: 103,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01332388 CAS Number: 2682-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...