2-Amino-N,N-dimethylbenzamide 95%
Supplier: Apollo Scientific
OR33559-1G
OR33559-5G
APOSOR33559-1GEA
176
EUR
APOSOR33559-1G
APOSOR33559-5G
2-Amino-N,N-dimethylbenzamide 95%
2-Amino-N,N-dimethylbenzamide
Formula:
C₉H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD01168906 CAS Number: 6526-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...