2-Chloro-4-piperidinobenzenecarbohydrazide 95%

Supplier: Apollo Scientific

OR33570-1G OR33570-5G
APOSOR33570-1GEA 336 EUR
APOSOR33570-1G APOSOR33570-5G
2-Chloro-4-piperidinobenzenecarbohydrazide 95%
2-Chloro-4-piperidinobenzenecarbohydrazide
Formula: C₁₂H₁₆ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD09607913
CAS Number: 952183-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR