2-Chloronaphthalene-1,3-dicarbaldehyde 95%

Supplier: Apollo Scientific

OR33593-1G
APOSOR33593-1GEA 270 EUR
APOSOR33593-1G
2-Chloronaphthalene-1,3-dicarbaldehyde 95%
2-Chloronaphthalene-1,3-dicarbaldehyde
Formula: C₁₂H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD18803523
CAS Number: 132554-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR