2-Formyl-5-picoline 95%
Supplier: Apollo Scientific
Synonyms:
5-Methylpyridine-2-carbaldehyde, 5-Methylpicolinaldehyde
OR33604-1G
OR33604-5G
APOSOR33604-1GEA
53.9
EUR
APOSOR33604-1G
APOSOR33604-5G
2-Formyl-5-picoline 95%
2-Formyl-5-picoline
Formula:
C₇H₇NO MW: 121,1400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07367942 CAS Number: 4985-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...