2-(Piperazin-1-yl)pyrazine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-(2-Pyrazinyl)piperazine
OR3374-1G
OR3374-500MG
OR3374-5G
APOSOR3374-1GEA
55.1
EUR
APOSOR3374-1G
APOSOR3374-500MG
APOSOR3374-5G
2-(Piperazin-1-yl)pyrazine 98%
2-(Piperazin-1-yl)pyrazine
Formula:
C₈H₁₂N₄ Boiling Pt: 105…108 °C/0,5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00040766 CAS Number: 34803-68-4 EINECS: 252-221-9 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...