2-(4-Pyridyl)malondialdehyde 95+%

Supplier: Apollo Scientific
Danger

Synonyms: 2-(Pyridin-4-yl)malonaldehyde

OR3395-1G OR3395-5G OR3395-500MG
APOSOR3395-1GEA 74.2 EUR
APOSOR3395-1G APOSOR3395-5G APOSOR3395-500MG
2-(4-Pyridyl)malondialdehyde 95+%
2-(4-Pyridyl)malondialdehyde
Formula: C₈H₇NO₂
MW: 149,149 g/mol
Melting Pt: 320 °C
Storage Temperature: Ambient
MDL Number: MFCD00216535
CAS Number: 51076-46-1
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR