2-Carboxythiophene-4-boronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR3433-1G
OR3433-5G
APOSOR3433-1GEA
364
EUR
APOSOR3433-1G
APOSOR3433-5G
2-Carboxythiophene-4-boronic acid 98%
2-Carboxythiophene-4-boronic acid
Formula:
C₅H₅BO₄S MW: 171,96 g/mol Melting Pt: 170…172 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08689521 CAS Number: 913835-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...