2,6-Dichloro-4-nitropyridine 97%

Supplier: Apollo Scientific
Warning

OR345059-25G OR345059-1G OR345059-5G
APOSOR345059-25GEA 82.8 EUR
APOSOR345059-25G APOSOR345059-1G APOSOR345059-5G
2,6-Dichloro-4-nitropyridine 97%
2,6-Dichloro-4-nitropyridine
Formula: C₅H₂Cl₂N₂O₂
MW: 192,99 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD05670545
CAS Number: 25194-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR