3-Amino-o-anisic acid 96%
Supplier: Apollo Scientific
|
Warning
|
OR345431-1G
OR345431-25G
OR345431-5G
APOSOR345431-1GEA
72.7
EUR
APOSOR345431-1G
APOSOR345431-25G
APOSOR345431-5G
3-Amino-o-anisic acid 96%
3-Amino-o-anisic acid
Formula:
C₈H₉NO₃ MW: 167,16 g/mol Melting Pt: 98…102 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09701221 CAS Number: 861306-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...