N'-[1-Amino-1-naphthalen-2-ylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
OR346013-1G
OR346013-500MG
APOSOR346013-1GEA
0
EUR
APOSOR346013-1G
APOSOR346013-500MG
N'-[1-Amino-1-naphthalen-2-ylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-naphthalen-2-ylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formula:
C₁₆H₁₉N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD10568193 CAS Number: 1053655-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...