Ethyl-5-(2-tert-butyloxycarbonylaminoethyl)[1,2,4]oxadiazole-3-carboxylate

Supplier: Apollo Scientific

OR346023-1G OR346023-500MG
APOSOR346023-1GEA 0 EUR
APOSOR346023-1G APOSOR346023-500MG
Ethyl-5-(2-tert-butyloxycarbonylaminoethyl)[1,2,4]oxadiazole-3-carboxylate
Ethyl-5-(2-tert-butyloxycarbonylaminoethyl)[1,2,4]oxadiazole-3-carboxylate
Formula: C₁₂H₁₉N₃O₅
MW: 285,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568205
CAS Number: 652158-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR