N'-[1-Cyclopentylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester

Supplier: Apollo Scientific

OR346031-1G OR346031-500MG
APOSOR346031-1GEA 0 EUR
APOSOR346031-1G APOSOR346031-500MG
N'-[1-Cyclopentylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Cyclopentylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
Formula: C₁₃H₂₅N₃O₂
MW: 255,36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568242
CAS Number: 1053657-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR