tert-Butyl-3-(aminomethyl)benzoate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
tert-Butyl 3-(aminomethyl)benzoate
OR346066-1G
APOSOR346066-1GEA
243
EUR
APOSOR346066-1G
tert-Butyl-3-(aminomethyl)benzoate
tert-Butyl-3-(aminomethyl)benzoate
Formula:
C₁₂H₁₇NO₂ MW: 207,27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08275208 CAS Number: 102638-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...