C-(3-Phenyl-[1,2,4]oxadiazol-5-yl)methylamine

Supplier: Apollo Scientific

Synonyms: 5-Aminomethyl-3-(phenyl)-[1,2,4]oxadiazole hydrochloride

OR346073-1G OR346073-5G
APOSOR346073-1GEA 0 EUR
APOSOR346073-1G APOSOR346073-5G
C-(3-Phenyl-[1,2,4]oxadiazol-5-yl)methylamine
C-(3-Phenyl-[1,2,4]oxadiazol-5-yl)methylamine
Formula: C₉H₉N₃O
MW: 175,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06166274
CAS Number: 90564-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR