Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate

Supplier: Apollo Scientific

Synonyms: Methyl 2-(((benzyloxy)carbonyl)amino)-3-hydroxypropanoate

OR346093-5G OR346093-25G
APOSOR346093-5GEA 0 EUR
APOSOR346093-5G APOSOR346093-25G
Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate
Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate
Formula: C₁₂H₁₅NO₅
MW: 253,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00136653
CAS Number: 14464-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR