5-Bromo-2-chloromethyl-1H-benzoimidazole
Supplier: Apollo Scientific
Synonyms:
5-Bromo-2-(chloromethyl)-1H-benzo[d]imidazole
OR346101-1G
OR346101-5G
OR346101-10G
APOSOR346101-1GEA
154
EUR
APOSOR346101-1G
APOSOR346101-5G
APOSOR346101-10G
5-Bromo-2-chloromethyl-1H-benzoimidazole
5-Bromo-2-chloromethyl-1H-benzoimidazole
Formula:
C₈H₆BrClN₂ MW: 245,51 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00457491 CAS Number: 1740-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...