2-Chloropyridine-3-sulphonic acid amide
Supplier: Apollo Scientific
|
Warning
|
OR346131-1G
OR346131-5G
APOSOR346131-1GEA
198
EUR
APOSOR346131-1G
APOSOR346131-5G
2-Chloropyridine-3-sulphonic acid amide
2-Chloropyridine-3-sulphonic acid amide
Formula:
C₅H₅ClN₂O₂S MW: 192,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08752623 CAS Number: 38025-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...