3-Methylisothiazol-5-ylamine hydrobromide
Supplier: Apollo Scientific
OR346139-1G
OR346139-5G
APOSOR346139-1GEA
0
EUR
APOSOR346139-1G
APOSOR346139-5G
3-Methylisothiazol-5-ylamine hydrobromide
3-Methylisothiazol-5-ylamine hydrobromide
Formula:
C₄H₇BrN₂S Storage Temperature: Ambient |
MDL Number:
MFCD12131107 CAS Number: 1208081-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...