(S)-1-(2-Chloro-acetyl)-pyrrolidine-2-carboxylic acid, Technical Grade

Supplier: Apollo Scientific
Warning

OR346168-1G OR346168-5G
APOSOR346168-1GEA 0 EUR
APOSOR346168-1G APOSOR346168-5G
(S)-1-(2-Chloro-acetyl)-pyrrolidine-2-carboxylic acid, Technical Grade
(S)-1-(2-Chloro-acetyl)-pyrrolidine-2-carboxylic acid
Formula: C₇H₁₀ClNO₃
Melting Pt: 112…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00046089
CAS Number: 23500-10-9
EINECS: 245-695-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR