5-(4-Bromophenyl)-3-methylisoxazole-4-carboxylic acid
Supplier: Apollo Scientific
OR346232-1G
OR346232-5G
APOSOR346232-1GEA
119
EUR
APOSOR346232-1G
APOSOR346232-5G
5-(4-Bromophenyl)-3-methylisoxazole-4-carboxylic acid
5-(4-Bromophenyl)-3-methylisoxazole-4-carboxylic acid
Formula:
C₁₁H₈BrNO₃ MW: 282,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16447083 CAS Number: 91182-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...