Ethyl-5-methyl-1,3,4-oxadiazole-2-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 5-methyl-1,3,4-oxadiazole-2-carboxylate, 5-Methyl-[1,3,4]oxadiazole-2-carboxylic acid ethyl ester

OR346245-1G OR346245-5G
APOSOR346245-1GEA 126 EUR
APOSOR346245-1G APOSOR346245-5G
Ethyl-5-methyl-1,3,4-oxadiazole-2-carboxylate
Ethyl-5-methyl-1,3,4-oxadiazole-2-carboxylate
Formula: C₆H₈N₂O₃
MW: 156,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752603
CAS Number: 37641-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR