Ethyl 3-Oxo-3-(4-chlorophenyl)propionate
Supplier: Apollo Scientific
Synonyms:
Ethyl 3-(4-chlorophenyl)-3-oxo-propionate, 3-Oxo-3-(4-chlorophenyl)propionic acid Ethyl ester
OR346250-1G
OR346250-5G
OR346250-10G
APOSOR346250-1GEA
55.3
EUR
APOSOR346250-1G
APOSOR346250-5G
APOSOR346250-10G
Ethyl 3-Oxo-3-(4-chlorophenyl)propionate
Ethyl 3-Oxo-3-(4-chlorophenyl)propionate
Formula:
C₁₁H₁₁ClO₃ MW: 226,66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00018713 CAS Number: 2881-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...