(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester

Supplier: Apollo Scientific

Synonyms: tert-Butyl (6-chloropyridin-3-yl)carbamate

OR346263-5G OR346263-1G OR346263-10G
APOSOR346263-5GEA 78.6 EUR
APOSOR346263-5G APOSOR346263-1G APOSOR346263-10G
(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester
(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester
Formula: C₁₀H₁₃ClN₂O₂
MW: 228,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062421
CAS Number: 171178-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR