5-Amino-3-methyl-1H-pyrazole-4-carbonitrile

Supplier: Apollo Scientific
Warning

OR346302-1G OR346302-5G
APOSOR346302-1GEA 161 EUR
APOSOR346302-1G APOSOR346302-5G
5-Amino-3-methyl-1H-pyrazole-4-carbonitrile
5-Amino-3-methyl-1H-pyrazole-4-carbonitrile
Formula: C₅H₆N₄
Melting Pt: 162…163 °C
Storage Temperature: Ambient
MDL Number: MFCD02020550
CAS Number: 5453-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR