4-Phenyl-1,2,3-thiadiazole 95+%
Supplier: Apollo Scientific
OR346388-1G
OR346388-5G
APOSOR346388-1GEA
154
EUR
APOSOR346388-1G
APOSOR346388-5G
4-Phenyl-1,2,3-thiadiazole 95+%
4-Phenyl-1,2,3-thiadiazole
Formula:
C₈H₆N₂S MW: 162,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00087232 CAS Number: 25445-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...