(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile 95+%

Supplier: Apollo Scientific

Synonyms: (S)-1-(2-Chloroacetyl)pyrrolidine-2-carbonitrile, (S)-1-(Chloroacetyl)-2-cyanopyrrolidine

OR346401-1G OR346401-5G
APOSOR346401-1GEA 49.5 EUR
APOSOR346401-1G APOSOR346401-5G
(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile 95+%
(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile
Formula: C₇H₉ClN₂O
MW: 172,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689902
CAS Number: 207557-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR