2-Chloro-N,N-dimethylnicotinamide 95+%
Supplier: Apollo Scientific
OR346435-1G
OR346435-5G
APOSOR346435-1GEA
0
EUR
APOSOR346435-1G
APOSOR346435-5G
2-Chloro-N,N-dimethylnicotinamide 95+%
2-Chloro-N,N-dimethylnicotinamide
Formula:
C₈H₉ClN₂O MW: 184.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08593346 CAS Number: 52943-21-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...