1-(4-Chlorophenyl)-1H-pyrazole-4-carbaldehyde 95+%
Supplier: Apollo Scientific
OR346441-1G
OR346441-5G
APOSOR346441-1GEA
201
EUR
APOSOR346441-1G
APOSOR346441-5G
1-(4-Chlorophenyl)-1H-pyrazole-4-carbaldehyde 95+%
1-(4-Chlorophenyl)-1H-pyrazole-4-carbaldehyde
Formula:
C₁₀H₇ClN₂O MW: 206,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07643175 CAS Number: 63874-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...