3-Chloro-1-methoxyisoquinoline 95+%

Supplier: Apollo Scientific

OR346496-1G
APOSOR346496-1GEA 166 EUR
APOSOR346496-1G
3-Chloro-1-methoxyisoquinoline 95+%
3-Chloro-1-methoxyisoquinoline
Formula: C₁₀H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD00160677
CAS Number: 24649-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR