4-Bromofuran-2-carboxamide 95+%

Supplier: Apollo Scientific

OR346577-1G OR346577-5G
APOSOR346577-1GEA 201 EUR
APOSOR346577-1G APOSOR346577-5G
4-Bromofuran-2-carboxamide 95+%
4-Bromofuran-2-carboxamide
Formula: C₅H₄BrNO
MW: 189,99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12755858
CAS Number: 957345-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR