3-Bromo-4-methoxybenzamide 95+%
Supplier: Apollo Scientific
OR346691-1G
APOSOR346691-1GEA
0
EUR
APOSOR346691-1G
3-Bromo-4-methoxybenzamide 95+%
3-Bromo-4-methoxybenzamide
Formula:
C₈H₈BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11643536 CAS Number: 200956-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...