2-Methyl-3-furoyl chloride 95+%
Supplier: Apollo Scientific
Synonyms:
2-Methylfuran-3-carbonyl chloride
OR346695-1G
OR346695-5G
APOSOR346695-1GEA
154
EUR
APOSOR346695-1G
APOSOR346695-5G
2-Methyl-3-furoyl chloride 95+%
2-Methyl-3-furoyl chloride
Formula:
C₆H₅ClO₂ Boiling Pt: 59…61 °C/12mmHg Density: 1,239 g/cm³ (25 °C) Flash Pt: 76 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03421418 CAS Number: 5555-00-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...